5-Hydroxy-2-adamantanone ≥98.0%

Supplier: TCI America

Synonyms: Idramantone, 5-Hydroxyadamantan-2-one

H0962-5G H0962-25G H0962-1G
TCH0962-5GEA 214.88 CAD
TCH0962-5G TCH0962-25G TCH0962-001G
5-Hydroxy-2-adamantanone ≥98.0%
5-Hydroxy-2-adamantanone

CAS Number: 20098-14-0
MDL Number: MFCD00192211
Molecular Formula: C10H14O2
Molecular Weight: 166.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
MDL Number: MFCD00192211
CAS Number: 20098-14-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93419bc4c0a24ce11b4f6f4fd6dab1597a1fb3e3&vid=93419bc4c0a24ce11b4f6f4fd6dab1597a1fb3e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR