4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0960-250G H0960-25G
TCH0960-250GEA 854.62 CAD
TCH0960-250G TCH0960-025G
4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)
4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture)

CAS Number: 17419-81-7
MDL Number: MFCD00077974
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal

Formula: C₇H₁₂O₃
MDL Number: MFCD00077974
CAS Number: 17419-81-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24e6f5af506a6d75760d8d952e2531c5706d620d&vid=24e6f5af506a6d75760d8d952e2531c5706d620d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR