1,2:5,6-Diepoxyhexane ≥96.0%
Supplier: TCI America
Synonyms:
1,5-Hexadiene diepoxide
H0953-1G
H0953-25G
H0953-5G
TCH0953-001GEA
80.58
CAD
TCH0953-001G
TCH0953-25G
TCH0953-5G
1,2:5,6-Diepoxyhexane ≥96.0%
1,2:5,6-Diepoxyhexane
CAS Number: 1888-89-7
MDL Number: MFCD01723447
Molecular Formula: C6H10O2
Molecular Weight: 114.14
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 188
Specific Gravity (20/20): 1.03
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2:5,6-Diepoxyhexane
Formula:
C₆H₁₀O₂ MW: 114.1424 g/mol Boiling Pt: 188 °C Density: 1.03 (20 °C) |
MDL Number:
MFCD01723447 CAS Number: 1888-89-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...