(R)-(+)-ɑ-Hydroxy-γ-butyrolactone ≥98.0%

Supplier: TCI America

H0950-1G H0950-100MG
TCH0950-001GEA 2809.24 CAD
TCH0950-001G TCH0950-100MG
(R)-(+)-ɑ-Hydroxy-γ-butyrolactone ≥98.0%
(R)-(+)-alpha-Hydroxy-gamma-butyrolactone

CAS Number: 56881-90-4
MDL Number: MFCD00211246
Molecular Formula: C4H6O3
Molecular Weight: 102.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 133
Flash Point (°C): 110
Specific Gravity (20/20): 1.32
Specific rotation [a]20/D: 71.5 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(+)-Dihydro-3-hydroxy-2(3H)-furanone, (R)-(+)-3-Hydroxytetrahydrofuran-2-one

Formula: C₄H₆O₃
Boiling Pt: 133 °C
Density: 1.32 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00211246
CAS Number: 56881-90-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b13fb2db1f14b67f25ac0cc91e00db406db527f&vid=3b13fb2db1f14b67f25ac0cc91e00db406db527f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR