O-Allylhydroxylamine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
A1449-1G
A1449-5G
TCA1449-001GEA
133.83
CAD
TCA1449-001G
TCA1449-005G
O-Allylhydroxylamine hydrochloride ≥98.0% (by titrimetric analysis)
O-Allylhydroxylamine hydrochloride
CAS Number: 38945-21-0
MDL Number: MFCD00012957
Molecular Formula: C3H7NO
Molecular Weight: 109.55
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 170
Formula:
C₃H₇NO Melting Pt: 170 °C |
MDL Number:
MFCD00012957 CAS Number: 38945-21-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...