7-Hydroxy-5-methyl-2-methylthio[1,2,4]triazolo[1,5-a]pyrimidine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 5-Methyl-2-methylthio-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 7-Hydroxy-5-methyl-2-methylthio-s-triazolo[1,5-a]pyrimidine, 4-Hydroxy-6-methyl-2-methylmercapto-1,3,3a,7-tetraazaindene, 5-Methyl-2-(methylthio)-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol

H0940-25G H0940-5G
TCH0940-025GEA 350.44 CAD
TCH0940-025G TCH0940-5G
7-Hydroxy-5-methyl-2-methylthio[1,2,4]triazolo[1,5-a]pyrimidine ≥98.0% (by HPLC)
7-Hydroxy-5-methyl-2-methylthio[1,2,4]triazolo[1,5-a]pyrimidine

CAS Number: 40775-78-8
MDL Number: MFCD00186154
Molecular Formula: C7H8N4OS
Molecular Weight: 196.23
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 4-Hydroxy-6-methyl-2-methylmercapto-1,3,3a,7-tetraazaindene, 5-Methyl-2-methylthio-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 7-Hydroxy-5-methyl-2-methylthio-s-triazolo[1,5-a]pyrimidine

Formula: C₇H₈N₄OS
MW: 196.23 g/mol
MDL Number: MFCD00186154
CAS Number: 40775-78-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3b522b9fc53f303be72db8e938356cb7431e195&vid=b3b522b9fc53f303be72db8e938356cb7431e195&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR