4-Hydroxy-4-phenylpiperidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0925-5G H0925-25G
TCH0925-005GEA 132.25 CAD
TCH0925-005G TCH0925-025G
4-Hydroxy-4-phenylpiperidine ≥98.0% (by GC, titration analysis)
4-Hydroxy-4-phenylpiperidine

CAS Number: 40807-61-2
MDL Number: MFCD00006000
Molecular Formula: C11H15NO
Molecular Weight: 177.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 160


Other Product Names / Synonyms: 4-Phenyl-4-piperidinol

Formula: C₁₁H₁₅NO
Melting Pt: 160 °C
MDL Number: MFCD00006000
CAS Number: 40807-61-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=340fe598b6cbdda9230b57c964687dd6113af71b&vid=340fe598b6cbdda9230b57c964687dd6113af71b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR