2,3,6,7,10,11-Hexahydroxytriphenylene ≥95.0% (by HPLC)
Supplier: TCI America
Synonyms:
Triphenylene-2,3,6,7,10,11-hexaol
H0907-1G
H0907-5G
TCH0907-001GEA
227.84
CAD
TCH0907-001G
TCH0907-005G
2,3,6,7,10,11-Hexahydroxytriphenylene ≥95.0% (by HPLC)
2,3,6,7,10,11-Hexahydroxytriphenylene
CAS Number: 4877-80-9
MDL Number: MFCD01321170
Molecular Formula: C18H12O6
Molecular Weight: 324.29
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Formula:
C₁₈H₁₂O₆ MW: 324.29 g/mol |
MDL Number:
MFCD01321170 CAS Number: 4877-80-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...