Methyl-3-hydroxy-2,2-dimethylpropanoate ≥98.0%
Supplier: TCI America
Synonyms:
Hydroxypivalic acid methyll ester, Methyl 2,2-Dimethyl-3-hydroxypropionate, Methyl 3-hydroxy-2,2-dimethylpropanoate, Methyl Hydroxypivalate, Methyl 2,2-dimethyl-3-hydroxypropionate, 2,2-Dimethyl-3-hydroxypropionic acid methyll ester
CAS Number: 14002-80-3
MDL Number: MFCD00009707
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 178
Melting point (°C): -16
Flash Point (°C): 77
Specific Gravity (20/20): 1.04
Other Product Names / Synonyms: 2,2-Dimethyl-3-hydroxypropionic Acid Methyl Ester, Hydroxypivalic Acid Methyl Ester, Methyl 2,2-Dimethyl-3-hydroxypropionate
Formula:
C₆H₁₂O₃ MW: 132.1600 g/mol Boiling Pt: 178 °C Melting Pt: –16 °C Density: 1.04 (20 °C) Flash Pt: 77 °C |
MDL Number:
MFCD00009707 CAS Number: 14002-80-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...