2-(tert-Butoxycarbonylamino)-1-ethanol ≥98.0%

Supplier: TCI America

Synonyms: N-(2-Hydroxyethyl)carbamic Acid tert-Butyl Ester, 2-(tert-Butoxycarbonyl)ethanolamine, N-(tert-Butoxycarbonyl)ethanolamine, 2-Boc-ethanolamine, tert-Butyl N-(2-Hydroxyethyl)carbamate, 2-(Boc-amino)-1-ethanol, Boc-Glycinol

H0899-5ML H0899-25ML
TCH0899-005MLEA 91.64 CAD
TCH0899-005ML TCH0899-025ML
2-(tert-Butoxycarbonylamino)-1-ethanol ≥98.0%
2-(tert-Butoxycarbonylamino)-1-ethanol

CAS Number: 26690-80-2
MDL Number: MFCD00056657
Molecular Formula: C7H15NO3
Molecular Weight: 161.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 109
Specific Gravity (20/20): 1.05


Other Product Names / Synonyms: 2-(Boc-amino)-1-ethanol, N-(2-Hydroxyethyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(2-Hydroxyethyl)carbamate, 2-(tert-Butoxycarbonyl)ethanolamine, 2-Boc-ethanolamine

Formula: C₇H₁₅NO₃
MW: 161.20 g/mol
Boiling Pt: 92 °C
Density: 1.05 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00056657
CAS Number: 26690-80-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be361e8ea551e0546b64321b908444ef34b9820e&vid=be361e8ea551e0546b64321b908444ef34b9820e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR