2,3,6,7,10,11-Hexamethoxytriphenylene ≥95.0%

Supplier: TCI America

H0898-1G H0898-5G
TCH0898-001GEA 230.68 CAD
TCH0898-001G TCH0898-005G
2,3,6,7,10,11-Hexamethoxytriphenylene ≥95.0%
2,3,6,7,10,11-Hexamethoxytriphenylene

CAS Number: 808-57-1
MDL Number: MFCD00075571
Molecular Formula: C24H24O6
Molecular Weight: 408.45
Purity/Analysis Method: >95.0% (GC)
Form: Crystal

Formula: C₂₄H₂₄O₆
MW: 408.45 g/mol
MDL Number: MFCD00075571
CAS Number: 808-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c64b2d724d5351a17c849260457362957111a4c6&vid=c64b2d724d5351a17c849260457362957111a4c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR