Ethyl-2-hydroxy-4-(trimethylsilyl)-3-butynoate ≥97.0%

Supplier: TCI America

H0895-1G H0895-5G
TCH0895-001GEA 218.2 CAD
TCH0895-001G TCH0895-5G
Ethyl-2-hydroxy-4-(trimethylsilyl)-3-butynoate ≥97.0%
Ethyl-2-hydroxy-4-(trimethylsilyl)-3-butynoate

CAS Number: 66697-09-4
MDL Number: MFCD00671544
Molecular Formula: C9H16O3Si
Molecular Weight: 200.31
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.98
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Hydroxy-4-(trimethylsilyl)-3-butynoic Acid Ethyl Ester

Formula: C₉H₁₆O₃Si
Density: 0.98 (20 °C)
MDL Number: MFCD00671544
CAS Number: 66697-09-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58300e1859ac74565f255a81ceb8881cf0c8634b&vid=58300e1859ac74565f255a81ceb8881cf0c8634b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR