4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate ≥97.0% free radical

Supplier: TCI America

H0878-1G H0878-5G
TCH0878-001GEA 121.66 CAD
TCH0878-001G TCH0878-005G
4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate ≥97.0% free radical
4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate

[Catalyst for Oxidation]
CAS Number: 3225-26-1
MDL Number: MFCD00075563
Molecular Formula: C16H22NO3
Molecular Weight: 276.36
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 104


Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Hydroxy-TEMPO Benzoate, 4-Benzoyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Benzoyloxy-TEMPO

Formula: C₁₆H₂₂NO₃
Melting Pt: 104 °C
MDL Number: MFCD00075563
CAS Number: 3225-26-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e809a1495c95c284c10115475f0f4e4a34c7240&vid=2e809a1495c95c284c10115475f0f4e4a34c7240&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR