8-Hydroxyquinoline-7-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0877-5G H0877-25G
TCH0877-005GEA 145.36 CAD
TCH0877-005G TCH0877-25G
8-Hydroxyquinoline-7-carboxylic acid ≥98.0% (by GC, titration analysis)
8-Hydroxyquinoline-7-carboxylic acid

CAS Number: 19829-79-9
MDL Number: MFCD00671539
Molecular Formula: C10H7NO3
Molecular Weight: 189.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal

Formula: C₁₀H₇NO₃
MDL Number: MFCD00671539
CAS Number: 19829-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9313aba49376bc488ac067c90818f2a345313bf8&vid=9313aba49376bc488ac067c90818f2a345313bf8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR