6-Hydroxypicolinic acid ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 6-Hydroxy-2-pyridinecarboxylic acid, 6-Hydroxypyridine-2-carboxylic acid

H0876-5G H0876-25G
TCH0876-005GEA 101.12 CAD
TCH0876-005G TCH0876-025G
6-Hydroxypicolinic acid ≥97.0% (by titrimetric analysis)
6-Hydroxypicolinic acid

CAS Number: 19621-92-2
MDL Number: MFCD00192220
Molecular Formula: C6H5NO3
Molecular Weight: 139.11
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Other Product Names / Synonyms: 6-Hydroxypicolinic Acid

Formula: C₆H₅NO₃
MW: 139.11 g/mol
MDL Number: MFCD00192220
CAS Number: 19621-92-2
EINECS: 417-690-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccdf386dd098c81c5f94115c1272c20df15abb37&vid=3c5fe168e2d2251b1c08ce72e03a7b5dc33dbc5c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR