4'-Hydroxyheptanophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0864-500G H0864-25G
TCH0864-500GEA 886.38 CAD
TCH0864-500G TCH0864-025G
4'-Hydroxyheptanophenone ≥98.0% (by GC, titration analysis)
4'-Hydroxyheptanophenone

CAS Number: 14392-72-4
MDL Number: MFCD00191523
Molecular Formula: C13H18O2
Molecular Weight: 206.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 220
Melting point (°C): 93


Other Product Names / Synonyms: Hexyl 4-Hydroxyphenyl Ketone

Formula: C₁₃H₁₈O₂
Boiling Pt: 220 °C
Melting Pt: 93 °C
MDL Number: MFCD00191523
CAS Number: 14392-72-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c7c737965d9f480be08a19f8d3ece93649d35dd&vid=9c7c737965d9f480be08a19f8d3ece93649d35dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR