(1S,2S,5S)-(-)-2-Hydroxy-3-pinanone ≥98.0%
Supplier: TCI America
Synonyms:
(1S,2S,5S)-(-)-2-Hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one
H0863-1G
H0863-5G
TCH0863-001GEA
131.14
CAD
TCH0863-001G
TCH0863-005G
(1S,2S,5S)-(-)-2-Hydroxy-3-pinanone ≥98.0%
(1S,2S,5S)-(-)-2-Hydroxy-3-pinanone
CAS Number: 1845-25-6
MDL Number: MFCD00040556
Molecular Formula: C10H16O2
Molecular Weight: 168.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 106
Specific rotation [a]20/D: -33 deg (C=0.5, CHCl3)
Product Categories: Bioscience, Bicyclic Monoterpenes
Other Product Names / Synonyms: (1S,2S,5S)-(-)-2-Hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one
Formula:
C₁₀H₁₆O₂ MW: 168.24 g/mol Flash Pt: 106 °C |
MDL Number:
MFCD00040556 CAS Number: 1845-25-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...