4-(2-Hydroxyethoxy)benzaldehyde ≥98.0%

Supplier: TCI America

H0859-25G H0859-5G
TCH0859-025GEA 284.24 CAD
TCH0859-025G TCH0859-5G
4-(2-Hydroxyethoxy)benzaldehyde ≥98.0%
4-(2-Hydroxyethoxy)benzaldehyde

CAS Number: 22042-73-5
MDL Number: MFCD00191450
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 40
Storage Temperature: 0-10°C

Formula: C₉H₁₀O₃
MW: 166.1739 g/mol
Melting Pt: 40 °C
MDL Number: MFCD00191450
CAS Number: 22042-73-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6984d6f61d56bd0f0033aa6f3172acccf0074bb&vid=f6984d6f61d56bd0f0033aa6f3172acccf0074bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR