3-(2-Hydroxyethoxy)benzaldehyde ≥96.0%
Supplier: TCI America
H0858-25G
H0858-5G
TCH0858-025GEA
254.54
CAD
TCH0858-025G
TCH0858-5G
3-(2-Hydroxyethoxy)benzaldehyde ≥96.0%
3-(2-Hydroxyethoxy)benzaldehyde
CAS Number: 60345-97-3
MDL Number: MFCD00191448
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.20
Storage Temperature: 0-10°C
Formula:
C₉H₁₀O₃ MW: 166.170 g/mol Density: 1.20 (20 °C) |
MDL Number:
MFCD00191448 CAS Number: 60345-97-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...