2-(2-Hydroxyethoxy)benzaldehyde ≥96.0%

Supplier: TCI America

H0857-25G H0857-5G
TCH0857-025GEA 390.26 CAD
TCH0857-025G TCH0857-5G
2-(2-Hydroxyethoxy)benzaldehyde ≥96.0%
2-(2-Hydroxyethoxy)benzaldehyde

CAS Number: 22042-72-4
MDL Number: MFCD00191449
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 134
Melting point (°C): 45
Storage Temperature: 0-10°C

Formula: C₉H₁₀O₃
MW: 166.1739 g/mol
Boiling Pt: 134 °C
Melting Pt: 45 °C
MDL Number: MFCD00191449
CAS Number: 22042-72-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1fb31ce04cfb783fa0bff6f489d6b7b357ed146f&vid=1fb31ce04cfb783fa0bff6f489d6b7b357ed146f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR