Methyl-5-(hydroxymethyl)-1H-imidazole-4-carboxylate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-(Hydroxymethyl)-1H-imidazole-4-carboxylic Acid Methyl Ester

H0840-1G H0840-5G
TCH0840-001GEA 355.5 CAD
TCH0840-001G TCH0840-5G
Methyl-5-(hydroxymethyl)-1H-imidazole-4-carboxylate ≥98.0% (by titrimetric analysis)
Methyl-5-(hydroxymethyl)-1H-imidazole-4-carboxylate

CAS Number: 82032-43-7
MDL Number: MFCD00191520
Molecular Formula: C6H8N2O3
Molecular Weight: 156.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 173


Other Product Names / Synonyms: 5-(Hydroxymethyl)-1H-imidazole-4-carboxylic Acid Methyl Ester

Formula: C₆H₈N₂O₃
MW: 156.14 g/mol
Melting Pt: 173 °C
MDL Number: MFCD00191520
CAS Number: 82032-43-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6236807d1b93e7ddcd0beb4177bd09f21dc894a8&vid=6236807d1b93e7ddcd0beb4177bd09f21dc894a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR