1-Hydroxy-2-naphthaldehyde ≥98.0%

Supplier: TCI America

H0839-1G H0839-5G
TCH0839-001GEA 180.12 CAD
TCH0839-001G TCH0839-005G
1-Hydroxy-2-naphthaldehyde ≥98.0%
1-Hydroxy-2-naphthaldehyde

CAS Number: 574-96-9
MDL Number: MFCD00092325
Molecular Formula: C11H8O2
Molecular Weight: 172.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 56


Other Product Names / Synonyms: 1-Naphthol-2-carboxaldehyde

Formula: C₁₁H₈O₂
MW: 172.1800 g/mol
Melting Pt: 56 °C
MDL Number: MFCD00092325
CAS Number: 574-96-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=237155ffd656f15688c3822c55deba55af00e248&vid=237155ffd656f15688c3822c55deba55af00e248&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR