Methyl-9-hydroxynonanoate ≥95.0%

Supplier: TCI America

H0835-5G H0835-1G
TCH0835-005GEA 1216.6 CAD
TCH0835-005G TCH0835-001G
Methyl-9-hydroxynonanoate ≥95.0%
Methyl-9-hydroxynonanoate

CAS Number: 34957-73-8
MDL Number: MFCD00191518
Molecular Formula: C10H20O3
Molecular Weight: 188.27
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 272
Specific Gravity (20/20): 0.97


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: 9-Hydroxypelargonic Acid Methyl Ester, 9-Hydroxynonanoic Acid Methyl Ester, Methyl 9-Hydroxypelargonate

Formula: C₁₀H₂₀O₃
Boiling Pt: 272 °C
Density: 0.97 (20 °C)
MDL Number: MFCD00191518
CAS Number: 34957-73-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70e7a08c8a3d09120c446e95e936e4c476f6f854&vid=70e7a08c8a3d09120c446e95e936e4c476f6f854&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR