Methyl (4-hydroxybenzoyl)acetate ≥93.0%

Supplier: TCI America

H0828-5G H0828-1G
TCH0828-005GEA 1357.06 CAD
TCH0828-005G TCH0828-001G
Methyl (4-hydroxybenzoyl)acetate ≥93.0%
Methyl (4-hydroxybenzoyl)acetate

CAS Number: 32066-29-8
MDL Number: MFCD00191516
Molecular Formula: C10H10O4
Molecular Weight: 194.19
Purity/Analysis Method: >93.0% (GC)
Form: Crystal
Melting point (°C): 90


Other Product Names / Synonyms: (4-Hydroxybenzoyl)acetic Acid Methyl Ester, Methyl 3-(4-Hydroxyphenyl)-3-oxopropionate, 3-(4-Hydroxyphenyl)-3-oxopropionic Acid Methyl Ester

Formula: C₁₀H₁₀O₄
Melting Pt: 90 °C
MDL Number: MFCD00191516
CAS Number: 32066-29-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f0cab7dd87834f4c09e51433c3db35d720af22a&vid=6f0cab7dd87834f4c09e51433c3db35d720af22a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR