1-(2-Hydroxyethyl)pyrrole ≥99.0%

Supplier: TCI America

H0826-1G H0826-5G
TCH0826-001GEA 175.22 CAD
TCH0826-001G TCH0826-005G
1-(2-Hydroxyethyl)pyrrole ≥99.0%
1-(2-Hydroxyethyl)pyrrole

CAS Number: 6719-02-4
MDL Number: MFCD00191451
Molecular Formula: C6H9NO
Molecular Weight: 111.14
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 215
Specific Gravity (20/20): 1.06

Formula: C₆H₉NO
Boiling Pt: 215 °C
Density: 1.06 (20 °C)
MDL Number: MFCD00191451
CAS Number: 6719-02-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36c081097c2ff8f87a8c62d286774bb08c701449&vid=36c081097c2ff8f87a8c62d286774bb08c701449&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR