4-Methoxyphenyl-4-hydroxybenzoate 99,0

Supplier: TCI America

Synonyms: 4-Methoxyphenylparaben, 4-Hydroxybenzoic Acid 4-Methoxyphenyl Ester

H0825-5G H0825-25G
TCH0825-005GEA 153.73 CAD
TCH0825-005G TCH0825-25G
4-Methoxyphenyl-4-hydroxybenzoate 99,0
4-Methoxyphenyl-4-hydroxybenzoate

CAS Number: 50687-62-2
MDL Number: MFCD00191515
Molecular Formula: C14H12O4
Molecular Weight: 244.25
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 194


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-Hydroxybenzoic Acid 4-Methoxyphenyl Ester, 4-Methoxyphenylparaben

Formula: C₁₄H₁₂O₄
MW: 244.25 g/mol
Melting Pt: 194 °C
MDL Number: MFCD00191515
CAS Number: 50687-62-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55b436aabe6aee50a917986c572da3baa9c69df5&vid=55b436aabe6aee50a917986c572da3baa9c69df5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR