Dimethyl-1,8-anthracenedicarboxylate ≥95.0%

Supplier: TCI America

A1433-1G A1433-5G
TCA1433-001GEA 413.96 CAD
TCA1433-001G TCA1433-5G
Dimethyl-1,8-anthracenedicarboxylate ≥95.0%
Dimethyl-1,8-anthracenedicarboxylate

CAS Number: 93655-34-6
MDL Number: MFCD01321126
Molecular Formula: C18H14O4
Molecular Weight: 294.31
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 105


Other Product Names / Synonyms: 1,8-Anthracenedicarboxylic Acid Dimethyl Ester

Formula: C₁₈H₁₄O₄
Melting Pt: 105 °C
MDL Number: MFCD01321126
CAS Number: 93655-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05c3f86bbdaac9199df06aba2d46de3bb2cf9b47&vid=05c3f86bbdaac9199df06aba2d46de3bb2cf9b47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR