Methyl-4-(2-hydroxyethoxy)benzoate ≥98.0%

Supplier: TCI America

H0820-5G H0820-25G
TCH0820-005GEA 200.66 CAD
TCH0820-005G TCH0820-025G
Methyl-4-(2-hydroxyethoxy)benzoate ≥98.0%
Methyl-4-(2-hydroxyethoxy)benzoate

CAS Number: 3204-73-7
MDL Number: MFCD00075872
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 66


Other Product Names / Synonyms: 4-(2-Hydroxyethoxy)benzoic Acid Methyl Ester

Formula: C₁₀H₁₂O₄
Melting Pt: 66 °C
MDL Number: MFCD00075872
CAS Number: 3204-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=34cfb4837ade9de82ddd8d5dcdede2cb5e60759d&vid=34cfb4837ade9de82ddd8d5dcdede2cb5e60759d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR