4-Cyanophenyl-4-heptylbenzoate ≥98.0%
Supplier: TCI America
H0810-5G
H0810-1G
TCH0810-5GEA
139.04
CAD
TCH0810-5G
TCH0810-001G
4-Cyanophenyl-4-heptylbenzoate ≥98.0%
4-Cyanophenyl 4-heptylbenzoate
CAS Number: 38690-76-5
MDL Number: MFCD00001817
Molecular Formula: C21H23NO2
Molecular Weight: 321.42
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 57
Flash Point (°C): 110
Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 4-Heptylbenzoic Acid 4-Cyanophenyl Ester
Formula:
C₂₁H₂₃NO₂ Melting Pt: 57 °C Flash Pt: 110 °C |
MDL Number:
MFCD00001817 CAS Number: 38690-76-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...