Hexaethylene glycol monomethyl ether ≥97.0%
Supplier: TCI America
Synonyms:
mPEG6-Alcohol, 2,5,8,11,14,17-Hexaoxanonadecan-19-ol
H0808-5G
H0808-25G
H0808-1G
TCH0808-5GEA
526.14
CAD
TCH0808-5G
TCH0808-25G
TCH0808-001G
Hexaethylene glycol monomethyl ether ≥97.0%
Hexaethylene glycol monomethyl ether
CAS Number: 23601-40-3
MDL Number: MFCD00045996
Molecular Formula: C13H28O7
Molecular Weight: 296.36
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 167
Specific Gravity (20/20): 1.08
Product Categories: Bioscience, PEGylation Reagents
Other Product Names / Synonyms: mPEG6-Alcohol
Formula:
C₁₃H₂₈O₇ Boiling Pt: 167 °C Density: 1.08 (20 °C) |
MDL Number:
MFCD00045996 CAS Number: 23601-40-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...