5-(4-Hydroxyphenyl)hydantoin ≥98.0%

Supplier: TCI America

Synonyms: 5-(4-Hydroxyphenyl)imidazolidine-2,4-dione

H0806-25G H0806-5G
TCH0806-025GEA 170.32 CAD
TCH0806-025G TCH0806-5G
5-(4-Hydroxyphenyl)hydantoin ≥98.0%
5-(4-Hydroxyphenyl)hydantoin

CAS Number: 2420-17-9
MDL Number: MFCD00044002
Molecular Formula: C9H8N2O3
Molecular Weight: 192.17
Purity/Analysis Method: >98.0% (N)
Form: Crystal

Formula: C₉H₈N₂O₃
MW: 192.17 g/mol
MDL Number: MFCD00044002
CAS Number: 2420-17-9

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR