2-(Carbobenzoxyamino)-1-ethanol ≥97.0%
Supplier: TCI America
H0793-5G
H0793-25G
TCH0793-005GEA
93.22
CAD
TCH0793-005G
TCH0793-025G
2-(Carbobenzoxyamino)-1-ethanol ≥97.0%
2-(Carbobenzoxyamino)-1-ethanol
CAS Number: 77987-49-6
MDL Number: MFCD00191060
Molecular Formula: C10H13NO3
Molecular Weight: 195.22
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 215
Melting point (°C): 60
Other Product Names / Synonyms: Benzyl N-(2-Hydroxyethyl)carbamate, 2-(Cbz-amino)-1-ethanol, N-(2-Hydroxyethyl)carbamic Acid Benzyl Ester
Formula:
C₁₀H₁₃NO₃ Boiling Pt: 215 °C Melting Pt: 60 °C |
MDL Number:
MFCD00191060 CAS Number: 77987-49-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...