ɑ-Hydroxy-γ-butyrolactone ≥98.0%

Supplier: TCI America

Synonyms: 3-Hydroxytetrahydrofuran-2-one, Dihydro-3-hydroxy-2(3H)-furanone, 3-Hydroxydihydrofuran-2(3H)-one

H0788-10G H0788-25G
TCH0788-010GEA 617.78 CAD
TCH0788-010G TCH0788-25G
ɑ-Hydroxy-γ-butyrolactone ≥98.0%
alpha-Hydroxy-gamma-butyrolactone

CAS Number: 19444-84-9
MDL Number: MFCD00134268
Molecular Formula: C4H6O3
Molecular Weight: 102.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 133
Specific Gravity (20/20): 1.33


Other Product Names / Synonyms: Dihydro-3-hydroxy-2(3H)-furanone, 3-Hydroxytetrahydrofuran-2-one

Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 133 °C
Density: 1.33 (20 °C)
MDL Number: MFCD00134268
CAS Number: 19444-84-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6bf0052eca4d5766be64f54e235d1caf88195a48&vid=6bf0052eca4d5766be64f54e235d1caf88195a48&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR