Dimethyl-trans-3-hexenedioate ≥98.0%
Supplier: TCI America
H0776-25G
H0776-5G
TCH0776-025GEA
543.52
CAD
TCH0776-025G
TCH0776-005G
Dimethyl-trans-3-hexenedioate ≥98.0%
Dimethyl-trans-3-hexenedioate
CAS Number: 25126-93-6
MDL Number: MFCD00145425
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Freezing point (°C): 17
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: trans-3-Hexenedioic Acid Dimethyl Ester
Formula:
C₈H₁₂O₄ Boiling Pt: 125 °C Density: 1.10 (20 °C) |
MDL Number:
MFCD00145425 CAS Number: 25126-93-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...