Sodium anthraquinone-1-sulfonate ≥98.0%

Supplier: TCI America

Synonyms: 1-Anthraquinonesulfonic acid sodium salt, Anthraquinone-1-sulphonic acid Sodium Salt, Sodium 9,10-dioxo-9,10-dihydroanthracene-1-sulfonate, Anthraquinone-1-sulfonic acid Sodium Salt, 9,10-Dihydro-9,10-dioxo-1-anthraquinonesulfonic acid sodium salt, 9,10-Dihydro-9,10-dioxo-1-anthraquinonesulphonic acid sodium salt, 1-Anthraquinonesulphonic acid sodium salt, Sodium 9,10-dioxo-9,10-dihydroanthracene-1-sulphonate

A1428-25G A1428-250G
TCA1428-025GEA 71.1 CAD
TCA1428-025G TCA1428-250G
Sodium anthraquinone-1-sulfonate ≥98.0%
Sodium anthraquinone-1-sulfonate

CAS Number: 128-56-3
MDL Number: MFCD00037145
Molecular Formula: C14H8O5S
Molecular Weight: 310.25
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: Anthraquinone-1-sulfonic Acid Sodium Salt

Formula: C₁₄H₈O₅S
MW: 310.25 g/mol
MDL Number: MFCD00037145
CAS Number: 128-56-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6949c58c7c2e871c9bea73867e30e4b7793eb92&vid=f6949c58c7c2e871c9bea73867e30e4b7793eb92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR