1-Hydroxy-4-nitro-9,10-anthraquinone ≥97.0% (by GC, titration analysis)
Supplier: TCI America
H0765-25G
H0765-5G
TCH0765-025GEA
704.68
CAD
TCH0765-025G
TCH0765-005G
1-Hydroxy-4-nitro-9,10-anthraquinone ≥97.0% (by GC, titration analysis)
1-Hydroxy-4-nitro-9,10-anthraquinone
CAS Number: 81-65-2
MDL Number: MFCD00142642
Molecular Formula: C14H7NO5
Molecular Weight: 269.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 272
Formula:
C₁₄H₇NO₅ Melting Pt: 272 °C |
MDL Number:
MFCD00142642 CAS Number: 81-65-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...