4-Hydroxy-2'-nitrobiphenyl ≥97.0%
Supplier: TCI America
H0753-1G
H0753-5G
TCH0753-001GEA
348.71
CAD
TCH0753-001G
TCH0753-5G
4-Hydroxy-2'-nitrobiphenyl ≥97.0%
4-Hydroxy-2'-nitrobiphenyl
CAS Number: 51264-59-6
MDL Number: MFCD00143326
Molecular Formula: C12H9NO3
Molecular Weight: 215.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 115
Other Product Names / Synonyms: 2'-Nitrobiphenyl-4-ol, 4-(2-Nitrophenyl)phenol
Formula:
C₁₂H₉NO₃ Melting Pt: 115 °C |
MDL Number:
MFCD00143326 CAS Number: 51264-59-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...