Allyl benzyl ether ≥99.0%

Supplier: TCI America

Synonyms: Allyl Benzyl Ether, ((Allyloxy)methyl)benzene

A1427-25ML A1427-100ML
TCA1427-025MLEA 147.1 CAD
TCA1427-025ML TCA1427-100ML
Allyl benzyl ether ≥99.0%
Allyl benzyl ether

CAS Number: 14593-43-2
MDL Number: MFCD00078288
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 76
Specific Gravity (20/20): 0.96

Formula: C₁₀H₁₂O
Boiling Pt: 205 °C
Density: 0.96 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00078288
CAS Number: 14593-43-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aba17467a2bbfdde5fdc81e9f2a6bead39f8c61d&vid=aba17467a2bbfdde5fdc81e9f2a6bead39f8c61d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR