2-(Hydroxymethyl)anthracene ≥98.0%
Supplier: TCI America
Synonyms:
2-Anthracenemethanol
H0750-5G
H0750-1G
TCH0750-5GEA
1317.72
CAD
TCH0750-5G
TCH0750-001G
2-(Hydroxymethyl)anthracene ≥98.0%
2-(Hydroxymethyl)anthracene
CAS Number: 22863-82-7
MDL Number: MFCD00142932
Molecular Formula: C15H12O
Molecular Weight: 208.26
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 224
Other Product Names / Synonyms: 2-Anthracenemethanol
Formula:
C₁₅H₁₂O Melting Pt: 224 °C |
MDL Number:
MFCD00142932 CAS Number: 22863-82-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...