4-Hydroxy-4'-nitrobiphenyl ≥98.0%
Supplier: TCI America
H0747-1G
H0747-5G
H0747-25G
TCH0747-001GEA
113.76
CAD
TCH0747-001G
TCH0747-005G
TCH0747-25G
4-Hydroxy-4'-nitrobiphenyl ≥98.0%
4-Hydroxy-4'-nitrobiphenyl
CAS Number: 3916-44-7
MDL Number: MFCD00047742
Molecular Formula: C12H9NO3
Molecular Weight: 215.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 207
Product Categories: Materials Chemistry, Organic Nonlinear Optical (NLO) Materials
Other Product Names / Synonyms: 4'-Nitrobiphenyl-4-ol
Formula:
C₁₂H₉NO₃ Melting Pt: 207 °C |
MDL Number:
MFCD00047742 CAS Number: 3916-44-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...