4-Hydroxyindanone ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-HYDROXY-2,3-DIHYDRO-1H-INDEN-1-ONE, 4-hydroxyindan-1-one, 4-Hydroxy-1-indanone
H0723-5G
H0723-25G
TCH0723-005GEA
1046.28
CAD
TCH0723-005G
TCH0723-025G
4-Hydroxyindanone ≥98.0% (by GC, titration analysis)
4-Hydroxyindanone
CAS Number: 40731-98-4
MDL Number: MFCD00143330
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Formula:
C₉H₈O₂ MW: 148.161 g/mol |
MDL Number:
MFCD00143330 CAS Number: 40731-98-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...