2-(5-tert-Butyl-2-hydroxyphenyl)benzotriazole ≥98.0%
Supplier: TCI America
H0719-25G
H0719-500G
TCH0719-025GEA
83.74
CAD
TCH0719-025G
TCH0719-500G
2-(5-tert-Butyl-2-hydroxyphenyl)benzotriazole ≥98.0%
2-(5-tert-Butyl-2-hydroxyphenyl)benzotriazole
CAS Number: 3147-76-0
MDL Number: MFCD03844795
Molecular Formula: C16H17N3O
Molecular Weight: 267.33
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 97
Formula:
C₁₆H₁₇N₃O MW: 267.33 g/mol Melting Pt: 97 °C |
MDL Number:
MFCD03844795 CAS Number: 3147-76-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...