2-(3,5-Di-tert-amyl-2-hydroxyphenyl)benzotriazole ≥98.0%

Supplier: TCI America

H0718-500G H0718-25G
TCH0718-500GEA 459.78 CAD
TCH0718-500G TCH0718-025G
2-(3,5-Di-tert-amyl-2-hydroxyphenyl)benzotriazole ≥98.0%
2-(3,5-Di-tert-amyl-2-hydroxyphenyl)benzotriazole

CAS Number: 25973-55-1
MDL Number: MFCD00142737
Molecular Formula: C22H29N3O
Molecular Weight: 351.49
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 82

Formula: C₂₂H₂₉N₃O
MW: 351.49 g/mol
Melting Pt: 82 °C
MDL Number: MFCD00142737
CAS Number: 25973-55-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b372cbffb3bf9e9f9a1e39ee446f4d4557a36a6&vid=3b372cbffb3bf9e9f9a1e39ee446f4d4557a36a6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR