2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-(2H-Benzo[d][1,2,3]triazol-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol

H0716-25G H0716-500G
TCH0716-025GEA 56.28 CAD
TCH0716-025G TCH0716-500G
2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole ≥98.0% (by HPLC, titration analysis)
2-(2-Hydroxy-5-tert-octylphenyl)benzotriazole

CAS Number: 3147-75-9
MDL Number: MFCD00013338
Molecular Formula: C20H25N3O
Molecular Weight: 323.44
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 104

Formula: C₂₀H₂₅N₃O
MW: 323.44 g/mol
Melting Pt: 104 °C
MDL Number: MFCD00013338
CAS Number: 3147-75-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36dfa0f7433edd9d280fe466d1af03dc21e0d860&vid=36dfa0f7433edd9d280fe466d1af03dc21e0d860&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR