Methyl (R)-(-)-3-hydroxyisobutyrate ≥99.0%

Supplier: TCI America

Synonyms: Methyl (R)-(-)-beta-hydroxyisobutyrate, (R)-Methyl 3-hydroxy-2-methylpropanoate

H0703-25G H0703-5G
TCH0703-25GEA 744.18 CAD
TCH0703-25G TCH0703-5G
Methyl (R)-(-)-3-hydroxyisobutyrate ≥99.0%
Methyl (R)-(-)-3-hydroxyisobutyrate

CAS Number: 72657-23-9
MDL Number: MFCD00063450
Molecular Formula: C5H10O3
Molecular Weight: 118.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): 81
Specific Gravity (20/20): 1.07
Specific rotation [a]20/D: -22.5 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: (R)-(-)-3-Hydroxyisobutyric Acid Methyl Ester, (R)-(-)-3-Hydroxy-2-methylpropionic Acid Methyl Ester, Methyl (R)-(-)-3-Hydroxy-2-methylpropionate

Formula: C₅H₁₀O₃
MW: 118.1311 g/mol
Boiling Pt: 60 °C
Density: 1.07 (20 °C)
Flash Pt: 81 °C
MDL Number: MFCD00063450
CAS Number: 72657-23-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c8ced2b7bcff02f73acf8c04a540955a2bd2c6e&vid=2c8ced2b7bcff02f73acf8c04a540955a2bd2c6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR