4-Hexyloxyphthalonitrile ≥95.0%

Supplier: TCI America

Synonyms: 1,2-Dicyano-4-hexyloxybenzene

H0691-1G H0691-5G
TCH0691-001GEA 158.63 CAD
TCH0691-001G TCH0691-005G
4-Hexyloxyphthalonitrile ≥95.0%
4-Hexyloxyphthalonitrile

CAS Number: 104949-82-8
MDL Number: MFCD00059077
Molecular Formula: C14H16N2O
Molecular Weight: 228.30
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.04


Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 1,2-Dicyano-4-hexyloxybenzene

Formula: C₁₄H₁₆N₂O
Density: 1.04 (20 °C)
MDL Number: MFCD00059077
CAS Number: 104949-82-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfb3fbffd6b5e4a20550d53001db9738d305ca9f&vid=dfb3fbffd6b5e4a20550d53001db9738d305ca9f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR