1-Acetyl-2-pyrrolidone ≥98.0%

Supplier: TCI America

A1418-5G A1418-25G
TCA1418-005GEA 143.78 CAD
TCA1418-005G TCA1418-025G
1-Acetyl-2-pyrrolidone ≥98.0%
1-Acetyl-2-pyrrolidone

CAS Number: 932-17-2
MDL Number: MFCD00014101
Molecular Formula: C6H9NO2
Molecular Weight: 127.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 231
Specific Gravity (20/20): 1.15

Formula: C₆H₉NO₂
Boiling Pt: 231 °C
Density: 1.15 (20 °C)
MDL Number: MFCD00014101
CAS Number: 932-17-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97338a70df77cad1fd87dbd8f14a34d124f857b8&vid=97338a70df77cad1fd87dbd8f14a34d124f857b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR