3-Indolepropanol ≥98.0%

Supplier: TCI America

H0680-1G H0680-5G
TCH0680-1GEA 173.48 CAD
TCH0680-1G TCH0680-5G
3-Indolepropanol ≥98.0%
3-Indolepropanol

CAS Number: 3569-21-9
MDL Number: MFCD00013437
Molecular Formula: C11H13NO
Molecular Weight: 175.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 200
Flash Point (°C): 110
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: 3-(3-Hydroxypropyl)indole, Homotryptophol

Formula: C₁₁H₁₃NO
Boiling Pt: 200 °C
Density: 1.13 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00013437
CAS Number: 3569-21-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=063fb2e86fba9285356df2d2b2b137e28882d6a4&vid=063fb2e86fba9285356df2d2b2b137e28882d6a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR