10,12-Heptacosadiynoic acid ≥98.0%
Supplier: TCI America
H0666-100MG
H0666-500MG
TCH0666-100MGEA
208.24
CAD
TCH0666-100MG
TCH0666-500MG
10,12-Heptacosadiynoic acid ≥98.0%
10,12-Heptacosadiynoic acid
CAS Number: 67071-94-7
MDL Number: MFCD00060065
Molecular Formula: C27H46O2
Molecular Weight: 402.66
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 69
Storage Temperature: 0-10°C
Formula:
C₂₇H₄₆O₂ Melting Pt: 69 °C |
MDL Number:
MFCD00060065 CAS Number: 67071-94-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...