2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-ɑ-D-glucopyranosyl chloride ≥93.0%

Supplier: TCI America

Synonyms: 1-Chloro-2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucose, 2-Acetamido-2-deoxy-α-D-glucopyranosyl chloride 3,4,6-triacetate, 2-Acetamido-2-deoxy-ɑ-D-glucopyranosyl Chloride 3,4,6-Triacetate, α-Acetochloro-D-glucosamine, 1-Chloro-2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-ɑ-D-glucose, (2R,3S,4R,5R,6R)-5-Acetamido-2-(acetoxymethyl)-6-chlorotetrahydro-2H-pyran-3,4-diyl diacetate, 2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-α-D-glucopyranosyl chloride

A1416-5G A1416-1G
TCA1416-005GEA 413.96 CAD
TCA1416-005G TCA1416-1G
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-ɑ-D-glucopyranosyl chloride ≥93.0%
2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-alpha-D-glucopyranosyl chloride

CAS Number: 3068-34-6
MDL Number: MFCD00069776
Molecular Formula: C14H20ClNO8
Molecular Weight: 365.76
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Specific rotation [a]20/D: 117.5 deg (C=1, CHCl3)
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Glycosylation, Bioscience, Halogenosugars, Glucosamine, Aminosugars, O-Substituted Sugars
Other Product Names / Synonyms: 2-Acetamido-2-deoxy-alpha-D-glucopyranosyl Chloride 3,4,6-Triacetate, 1-Chloro-2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-alpha-D-glucose

Formula: C₁₄H₂₀ClNO₈
MDL Number: MFCD00069776
CAS Number: 3068-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e2f97695063b4520afad6b34e98ce31607b8941&vid=8e2f97695063b4520afad6b34e98ce31607b8941&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR